4-Chloropicolinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C1360-1G C1360-5G
TCC1360-001GEA 70 USD
TCC1360-001G TCC1360-005G
4-Chloropicolinic acid ≥98.0% (by GC, titration analysis)
4-Chloropicolinic acid

CAS Number: 5470-22-4
MDL Number: MFCD00191400
Molecular Formula: C6H4ClNO2
Molecular Weight: 157.55
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 181


Other Product Names / Synonyms: 4-Chloropicolinic Acid

Formula: C₆H₄ClNO₂
Melting Pt: 181 °C
MDL Number: MFCD00191400
CAS Number: 5470-22-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6676fb5bf3d83cc7c44c6632faf7b7a26d0c037b&vid=6676fb5bf3d83cc7c44c6632faf7b7a26d0c037b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR