3-Thiophenecarbonitrile ≥98.0%

Supplier: TCI America

Synonyms: 3-Cyanothiophene, 3-Thienyl cyanide, Thiophene-3-carbonitrile

C1344-25G C1344-5G
TCC1344-25GEA 200.9 USD
TCC1344-25G TCC1344-005G
3-Thiophenecarbonitrile ≥98.0%
3-Thiophenecarbonitrile

CAS Number: 1641-09-4
MDL Number: MFCD00151852
Molecular Formula: C5H3NS
Molecular Weight: 109.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Specific Gravity (20/20): 1.19


Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 3-Thiophenecarbonitrile

Formula: C₅H₃NS
MW: 109.15 g/mol
Boiling Pt: 205 °C
Density: 1.19 (20 °C)
MDL Number: MFCD00151852
CAS Number: 1641-09-4
EINECS: 216-687-7
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de4c54e6dcbd6ca5dc02cf541042ed55ef61313c&vid=de4c54e6dcbd6ca5dc02cf541042ed55ef61313c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR