2'-Chloro-4'-fluoroacetanilide ≥98.0%

Supplier: TCI America

Synonyms: N-(2-Chloro-4-fluorophenyl)acetamide

C1311-25G C1311-5G
TCC1311-025GEA 148 USD
TCC1311-025G TCC1311-5G
2'-Chloro-4'-fluoroacetanilide ≥98.0%
2'-Chloro-4'-fluoroacetanilide

CAS Number: 399-35-9
MDL Number: MFCD00042594
Molecular Formula: C8H7ClFNO
Molecular Weight: 187.60
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 118

Formula: C₈H₇ClFNO
MW: 187.6 g/mol
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 118 °C
MDL Number: MFCD00042594
CAS Number: 399-35-9

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR