1-Chloro-3-methyl-2-butene ≥85.0% stabilized

Supplier: TCI America


Stabilization: Stabilised with Stabilised

C1290-250ML C1290-25ML
TCC1290-250MLEA 759 USD
TCC1290-250ML TCC1290-25ML
1-Chloro-3-methyl-2-butene ≥85.0% stabilized
1-Chloro-3-methyl-2-butene

(stabilized with K2CO3)
CAS Number: 503-60-6
MDL Number: MFCD00000982
Molecular Formula: C5H9Cl
Molecular Weight: 104.58
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Boiling point (°C): 109
Flash Point (°C): 15
Specific Gravity (20/20): 0.93


Other Product Names / Synonyms: Prenyl Chloride

Formula: C₅H₉Cl
MW: 104.58 g/mol
Boiling Pt: 109 °C
Density: 0.93 (20 °C)
Flash Pt: 15 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000982
CAS Number: 503-60-6
EINECS: 207-972-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=912c01e8301d0d33a6a775cf5f02ce6536ab5b6b&vid=06942cca8bfa03000debde50f91a196f3e96c17d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >85.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR