4-Chlorosalicylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Chloro-2-hydroxybenzoic acid
C1282-500G
C1282-25G
TCC1282-500GEA
628
USD
TCC1282-500G
TCC1282-025G
4-Chlorosalicylic acid ≥98.0% (by GC, titration analysis)
4-Chlorosalicylic acid
CAS Number: 5106-98-9
MDL Number: MFCD00002449
Molecular Formula: C7H5ClO3
Molecular Weight: 172.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 217
Other Product Names / Synonyms: 4-Chloro-2-hydroxybenzoic Acid
Formula:
C₇H₅ClO₃ MW: 172.57 g/mol Melting Pt: 217 °C |
MDL Number:
MFCD00002449 CAS Number: 5106-98-9 EINECS: 225-829-7 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...