tert-Butyl chloroacetate ≥97.0%

Supplier: TCI America

C1267-500ML C1267-25ML
TCC1267-500MLEA 95 USD
TCC1267-500ML TCC1267-025ML
tert-Butyl chloroacetate ≥97.0%
tert-Butyl chloroacetate

CAS Number: 107-59-5
MDL Number: MFCD00000930
Molecular Formula: C6H11ClO2
Molecular Weight: 150.60
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 60
Flash Point (°C): 28
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: Chloroacetic Acid tert-Butyl Ester

Formula: C₆H₁₁ClO₂
MW: 150.6 g/mol
Boiling Pt: 60 °C
Density: 1.05 (20 °C)
Flash Pt: 28 °C
MDL Number: MFCD00000930
CAS Number: 107-59-5
EINECS: 203-506-1
UN: 2929
ADR: 6.1,II
Merck Index: 13,01562
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc20e6cd3c9d4c39b6f7258f53745c8a17f2dfa1&vid=dc20e6cd3c9d4c39b6f7258f53745c8a17f2dfa1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR