6-Chloroindole ≥98.0%

Supplier: TCI America

Synonyms: 6-Chloro-1H-indole

C1249-1G C1249-5G
TCC1249-001GEA 30 USD
TCC1249-001G TCC1249-005G
6-Chloroindole ≥98.0%
6-Chloroindole

CAS Number: 17422-33-2
MDL Number: MFCD00005681
Molecular Formula: C8H6ClN
Molecular Weight: 151.59
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 89

Formula: C₈H₆ClN
MW: 151.6 g/mol
Melting Pt: 89 °C
MDL Number: MFCD00005681
CAS Number: 17422-33-2
EINECS: 241-449-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a915814fcba38cbc6619cef8d9764aad29ba8738&vid=a915814fcba38cbc6619cef8d9764aad29ba8738&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR