α,4-Dichloro-α-phenyltoluene ≥96.0%

Supplier: TCI America
Danger

Synonyms: 4-Chlorobenzhydryl chloride, 1-Chloro-4-(chloro(phenyl)methyl)benzene

C1234-500G C1234-25G
TCC1234-500GEA 313 USD
TCC1234-500G TCC1234-25G
α,4-Dichloro-α-phenyltoluene ≥96.0%
α,4-Dichloro-α-phenyltoluene

CAS Number: 134-83-8
MDL Number: MFCD00000856
Molecular Formula: C13H10Cl2
Molecular Weight: 237.12
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Flash Point (°C): 113
Specific Gravity (20/20): 1.24


Other Product Names / Synonyms: Chloro(4-chlorophenyl)phenylmethane

Formula: C₁₃H₁₀Cl₂
MW: 237.13 g/mol
Boiling Pt: 160 °C
Density: 1.24 (20 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000856
CAS Number: 134-83-8
EINECS: 205-158-6
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b797b56134134bba9d7ceedb81e2d4ab23fe3b0&vid=9b797b56134134bba9d7ceedb81e2d4ab23fe3b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR