2-Chloro-5-nitrotoluene ≥98.0%
Supplier: TCI America
Synonyms:
1-Chloro-2-methyl-4-nitrobenzene
C1233-500G
C1233-25G
TCC1233-500GEA
413.2
USD
TCC1233-500G
TCC1233-25G
2-Chloro-5-nitrotoluene ≥98.0%
2-Chloro-5-nitrotoluene
CAS Number: 13290-74-9
MDL Number: MFCD00041430
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 96
Melting point (°C): 44
Other Product Names / Synonyms: 4-Chloro-3-methylnitrobenzene, 5-Nitro-2-chlorotoluene
Formula:
C₇H₆ClNO₂ Boiling Pt: 96 °C Melting Pt: 44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041430 CAS Number: 13290-74-9 EINECS: 236-306-8 UN: 3457 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...