2-Chloro-4-fluorotoluene ≥98.0%

Supplier: TCI America

C1227-500G C1227-25G
TCC1227-500GEA 275.1 USD
TCC1227-500G TCC1227-25G
2-Chloro-4-fluorotoluene ≥98.0%
2-Chloro-4-fluorotoluene

CAS Number: 452-73-3
MDL Number: MFCD00000572
Molecular Formula: C7H6ClF
Molecular Weight: 144.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 153
Flash Point (°C): 50
Specific Gravity (20/20): 1.20

Formula: C₇H₆ClF
Boiling Pt: 153 °C
Density: 1.20 (20 °C)
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00000572
CAS Number: 452-73-3
EINECS: 207-209-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6180a8e1a82536ff7fa5e899dce998d537af191d&vid=305421020c19226e472d1cc3ac8bdbeb2bbfcfcf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR