Chroman-4-one ≥98.0%
Supplier: TCI America
C1175-5G
C1175-25G
TCC1175-005GEA
55.4
USD
TCC1175-005G
TCC1175-025G
Chroman-4-one ≥98.0%
Chroman-4-one
CAS Number: 491-37-2
MDL Number: MFCD00006840
Molecular Formula: C9H8O2
Molecular Weight: 148.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 128
Flash Point (°C): 110
Freezing point (°C): 34
Storage Temperature: 0-10°C
Formula:
C₉H₈O₂ Boiling Pt: 128 °C Melting Pt: 39 °C Density: 1.1291 g/cm³ (99.8 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00006840 CAS Number: 491-37-2 EINECS: 207-736-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...