4-Chlorothioanisole ≥98.0%

Supplier: TCI America
Warning

Synonyms: 1-Chloro-4-(methylthio)benzene, 4-Chlorophenyl methyl sulphide

C1156-25G C1156-10G
TCC1156-25GEA 75 USD
TCC1156-25G TCC1156-010G
4-Chlorothioanisole ≥98.0%
4-Chlorothioanisole

CAS Number: 123-09-1
MDL Number: MFCD00013643
Molecular Formula: C7H7ClS
Molecular Weight: 158.64
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 224
Specific Gravity (20/20): 1.22


Other Product Names / Synonyms: 1-Chloro-4-(methylthio)benzene, 4-Chlorophenyl Methyl Sulfide

Formula: C₇H₇ClS
MW: 158.65 g/mol
Boiling Pt: 224 °C
Melting Pt: 27…28 °C
Density: 1.22 (20 °C)
MDL Number: MFCD00013643
CAS Number: 123-09-1
EINECS: 204-600-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85638ff184eb9713b5e5572d90c42974766ca902&vid=85638ff184eb9713b5e5572d90c42974766ca902&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR