4-Acetylbenzonitrile ≥97.0%

Supplier: TCI America

C1141-25G C1141-10G
TCC1141-025GEA 53.4 USD
TCC1141-025G TCC1141-010G
4-Acetylbenzonitrile ≥97.0%
4-Acetylbenzonitrile

CAS Number: 1443-80-7
MDL Number: MFCD00001825
Molecular Formula: C9H7NO
Molecular Weight: 145.16
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 96
Melting point (°C): 56


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Acetylbenzonitrile

Formula: C₉H₇NO
Boiling Pt: 96 °C
Melting Pt: 56 °C
MDL Number: MFCD00001825
CAS Number: 1443-80-7
EINECS: 215-885-0
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ed1343c3600dda45d186801b1ba654d38ce03f4&vid=7ed1343c3600dda45d186801b1ba654d38ce03f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR