5-Chloro-2-nitroaniline ≥99.0%

Supplier: TCI America
Danger

C1139-25G C1139-250G
TCC1139-25GEA 50 USD
TCC1139-25G TCC1139-250G
5-Chloro-2-nitroaniline ≥99.0%
5-Chloro-2-nitroaniline

CAS Number: 1635-61-6
MDL Number: MFCD00007776
Molecular Formula: C6H5ClN2O2
Molecular Weight: 172.57
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 128

Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00007776
CAS Number: 1635-61-6
EINECS: 216-661-5
UN: 2237
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=468c6b6d24b59ad53cb1849896dea97767e105aa&vid=18c48717a2281889dba347d36c9d79cea64a3dec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR