6-Chloronicotinic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 6-Chloropyridine-3-carboxylic acid, 6-Chloro-3-pyridinecarboxylic acid

C1116-25G C1116-100G
TCC1116-025GEA 86.8 USD
TCC1116-025G TCC1116-100G
6-Chloronicotinic acid ≥98.0% (by titrimetric analysis)
6-Chloronicotinic acid

CAS Number: 5326-23-8
MDL Number: MFCD00006241
Molecular Formula: C6H4ClNO2
Molecular Weight: 157.55
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 6-Chloro-3-pyridinecarboxylic Acid

Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: 188…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00006241
CAS Number: 5326-23-8
EINECS: 226-201-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c362a62664f042a2af724498af595a35f97d007a&vid=c362a62664f042a2af724498af595a35f97d007a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR