Ethyl carbazate ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Ethyl hydrazinecarboxylate

C1097-500G C1097-25G
TCC1097-500GEA 997 USD
TCC1097-500G TCC1097-025G
Ethyl carbazate ≥99.0% (by titrimetric analysis)
Ethyl carbazate

CAS Number: 4114-31-2
MDL Number: MFCD00007595
Molecular Formula: C3H8N2O2
Molecular Weight: 104.11
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Boiling point (°C): 118
Melting point (°C): 47
Flash Point (°C): 86


Other Product Names / Synonyms: Ethoxycarbonylhydrazine, Carbazic Acid Ethyl Ester

Formula: C₃H₈N₂O₂
MW: 104.11 g/mol
Boiling Pt: 118 °C
Melting Pt: 47 °C
Flash Pt: 86 °C
MDL Number: MFCD00007595
CAS Number: 4114-31-2
EINECS: 223-903-3
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5774f9936bc430922668d36e4283c27f1baf946&vid=f5774f9936bc430922668d36e4283c27f1baf946&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR