1-(2-Chloroethyl)piperidinium chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
C1073-500G
C1073-25G
TCC1073-500GEA
458
USD
TCC1073-500G
TCC1073-25G
1-(2-Chloroethyl)piperidinium chloride ≥98.0% (by titrimetric analysis)
1-(2-Chloroethyl)piperidinium chloride
CAS Number: 2008-75-5
MDL Number: MFCD00012837
Molecular Formula: C7H14ClN
Molecular Weight: 184.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 232
Other Product Names / Synonyms: 2-Piperidinoethyl Chloride Hydrochloride
Formula:
C₇H₁₅Cl₂N MW: 184.11 g/mol Melting Pt: 232 °C |
MDL Number:
MFCD00012837 CAS Number: 2008-75-5 EINECS: 217-920-5 UN: 2928 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...