Ethyl oxalyl chloride ≥98% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Ethyl oxalyl monochloride, Oxalic Acid Monoethyl Ester Chloride, Ethyl Chlorooxoacetate, Ethyl Chloroglyoxylate, Chloroglyoxylic Acid Ethyl Ester, Ethyl chloroformylformate

C1066-500G C1066-25G
TCC1066-500GEA 546 USD
TCC1066-500G TCC1066-25G
Ethyl oxalyl chloride ≥98% (by titrimetric analysis)
Ethyl oxalyl chloride

CAS Number: 4755-77-5
MDL Number: MFCD00000706
Molecular Formula: C4H5ClO3
Molecular Weight: 136.53
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 135
Flash Point (°C): 41
Specific Gravity (20/20): 1.23


Other Product Names / Synonyms: Chloroglyoxylic Acid Ethyl Ester, Ethyl Chlorooxoacetate, Ethyl Oxalyl Chloride, Oxalic Acid Monoethyl Ester Chloride

Formula: C₄H₅ClO₃
MW: 136.53 g/mol
MDL Number: MFCD00000706
CAS Number: 4755-77-5
EINECS: 225-285-0
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cc33659c5e69bce1ec23eb9b6e8664d04c7bbe4&vid=0cc33659c5e69bce1ec23eb9b6e8664d04c7bbe4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)
Boiling point (°C) 135
Flash Point (°C) 41
Specific Gravity (20/20) 1.23

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR