2-Chloro-1-fluoro-4-nitrobenzene ≥95.0%

Supplier: TCI America

Synonyms: 3-Chloro-4-fluoronitrobenzene, 1-Chloro-2-fluoro-5-nitrobenzene

C1041-25G C1041-250G
TCC1041-025GEA 16.28 USD
TCC1041-025G TCC1041-250G
2-Chloro-1-fluoro-4-nitrobenzene ≥95.0%
2-Chloro-1-fluoro-4-nitrobenzene

CAS Number: 350-30-1
MDL Number: MFCD00007206
Molecular Formula: C6H3ClFNO2
Molecular Weight: 175.54
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 105
Melting point (°C): 43
Flash Point (°C): 113
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Chloro-1-fluoro-4-nitrobenzene

Formula: C₆H₃ClFNO₂
MW: 175.55 g/mol
Boiling Pt: 105 °C
Melting Pt: 43 °C
Flash Pt: 113 °C
MDL Number: MFCD00007206
CAS Number: 350-30-1
EINECS: 206-499-3
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=424ca5a246e2d05b8492df20d0e456d9579369e2&vid=b55d8d84263d0f79de7323887295e55803816c14&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR