3-Chloro-4-fluoroaniline ≥98.0%

Supplier: TCI America
Warning

C1024-500G C1024-25G
TCC1024-500GEA 75 USD
TCC1024-500G TCC1024-25G
3-Chloro-4-fluoroaniline ≥98.0%
3-Chloro-4-fluoroaniline

CAS Number: 367-21-5
MDL Number: MFCD00007767
Molecular Formula: C6H5ClFN
Molecular Weight: 145.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 45
Flash Point (°C): 110

Formula: C₆H₅ClFN
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 45 °C
Density: 1.226 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007767
CAS Number: 367-21-5
EINECS: 206-682-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c61a94e44032ef9ca544fa3c698eb448348243e5&vid=bbb57b5589a626db8f280b27ecc414dc2acbd209&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR