Cyclopropylméthanol ≥98.0%
Supplier: TCI America
Synonyms:
Cyclopropyl carbinol, Cyclopropanemethanol
C0982-5ML
C0982-25ML
TCC0982-005MLEA
22
USD
TCC0982-005ML
TCC0982-025ML
Cyclopropylméthanol ≥98.0%
Cyclopropylméthanol
CAS Number: 2516-33-8
MDL Number: MFCD00001309
Molecular Formula: C4H8O
Molecular Weight: 72.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 123
Flash Point (°C): 35
Specific Gravity (20/20): 0.91
Other Product Names / Synonyms: Cyclopropylmethanol, (Hydroxymethyl)cyclopropane, Cyclopropylcarbinol
Formula:
C₄H₈O MW: 72.11 g/mol Boiling Pt: 123 °C Melting Pt: –60 °C Density: 0.91 (20 °C) Flash Pt: 35 °C |
MDL Number:
MFCD00001309 CAS Number: 2516-33-8 EINECS: 219-735-5 UN: 1987 ADR: 3 III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...