5-Chlorouracil ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-Chloropyrimidine-2,4(1H,3H)-dione, Chlorouracil

C0969-25G C0969-5G
TCC0969-025GEA 169.5 USD
TCC0969-025G TCC0969-5G
5-Chlorouracil ≥98.0% (by titrimetric analysis)
5-Chlorouracil

CAS Number: 1820-81-1
MDL Number: MFCD00006019
Molecular Formula: C4H3ClN2O2
Molecular Weight: 146.53
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₄H₃ClN₂O₂
MW: 146.53 g/mol
Melting Pt: ? 300 °C
MDL Number: MFCD00006019
CAS Number: 1820-81-1
EINECS: 217-339-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=155ccdc5237004981be8455c6c17fd6105b1657c&vid=155ccdc5237004981be8455c6c17fd6105b1657c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR