α-Chloro-o-xylene ≥98.0%

Supplier: TCI America

C0864-500G C0864-25G
TCC0864-500GEA 276.71 USD
TCC0864-500G TCC0864-25G
α-Chloro-o-xylene ≥98.0%
α-Chloro-o-xylene

CAS Number: 552-45-4
MDL Number: MFCD00000899
Molecular Formula: C8H9Cl
Molecular Weight: 140.61
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 198
Flash Point (°C): 97
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: 2-Methylbenzyl Chloride

Formula: C₈H₉Cl
Boiling Pt: 198 °C
Melting Pt: –2 °C
Density: 1.09 (20 °C)
Flash Pt: 97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000899
CAS Number: 552-45-4
EINECS: 209-013-8
UN: 3265
ADR: 8,II
Merck Index: 12,10224
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfe368a0d8d35319390a47b6b0a6ce80b909158a&vid=2969f3a34b0888435a097547f73cf45aea056e6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR