18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥98.0%

Supplier: TCI America
Warning

C0860-100G C0860-5G C0860-25G
TCC0860-100GEA 266 USD
TCC0860-100G TCC0860-005G TCC0860-025G
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥98.0%
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane)

CAS Number: 17455-13-9
MDL Number: MFCD00005113
Molecular Formula: C12H24O6
Molecular Weight: 264.32
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 190
Melting point (°C): 40
Flash Point (°C): 113
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 1,4,7,10,13,16-Hexaoxacyclooctadecane

Formula: C₁₂H₂₄O₆
Boiling Pt: 190 °C
Melting Pt: 40 °C
Density: 1.237 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00005113
CAS Number: 17455-13-9
EINECS: 241-473-5
Merck Index: 13,02630
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=863edf00c424d4bd528953c3f2d0942b1eef685b&vid=863edf00c424d4bd528953c3f2d0942b1eef685b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR