4-Chlorobutyryl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Chlorobutanoyl Chloride

C0824-500ML C0824-25ML
TCC0824-500MLEA 100 USD
TCC0824-500ML TCC0824-25ML
4-Chlorobutyryl chloride ≥98.0% (by GC, titration analysis)
4-Chlorobutyryl chloride

CAS Number: 4635-59-0
MDL Number: MFCD00000754
Molecular Formula: C4H6Cl2O
Molecular Weight: 140.99
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 174
Flash Point (°C): 72
Specific Gravity (20/20): 1.26

Formula: C₄H₆Cl₂O
MW: 141 g/mol
Boiling Pt: 174 °C
Density: 1.26 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00000754
CAS Number: 4635-59-0
EINECS: 225-059-1
UN: 2927
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=591fec92a52bafab80e43b9d13cce911e106f3bd&vid=591fec92a52bafab80e43b9d13cce911e106f3bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR