2,2,2-Trichloroethyl chloroformate ≥98.0%

Supplier: TCI America

C0795-250G C0795-25G
TCC0795-250GEA 841 USD
TCC0795-250G TCC0795-25G
2,2,2-Trichloroethyl chloroformate ≥98.0%
2,2,2-Trichloroethyl chloroformate

CAS Number: 17341-93-4
MDL Number: MFCD00000810
Molecular Formula: C3H2Cl4O2
Molecular Weight: 211.85
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 172
Specific Gravity (20/20): 1.58


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: Chloroformic Acid 2,2,2-Trichloroethyl Ester

Formula: C₃H₂Cl₄O₂
MW: 211.86 g/mol
Boiling Pt: 172 °C
Melting Pt: 1 °C
Density: 1.58 (20 °C)
MDL Number: MFCD00000810
CAS Number: 17341-93-4
EINECS: 241-363-7
UN: 3277
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2d10fa30225cd48ed02f6a524eabb6c9e8e3c1&vid=ee2d10fa30225cd48ed02f6a524eabb6c9e8e3c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR