2-Cyclohexen-1-one ≥96.0%

Supplier: TCI America

C0707-25ML C0707-100ML
TCC0707-25MLEA 43.53 USD
TCC0707-25ML TCC0707-100ML
2-Cyclohexen-1-one ≥96.0%
2-Cyclohexen-1-one

CAS Number: 930-68-7
MDL Number: MFCD00001577
Molecular Formula: C6H8O
Molecular Weight: 96.13
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 171
Flash Point (°C): 56
Specific Gravity (20/20): 1.00

Formula: C₆H₈O
MW: 96.13 g/mol
Boiling Pt: 171 °C
Density: 1.00 (20 °C)
Flash Pt: 56 °C
MDL Number: MFCD00001577
CAS Number: 930-68-7
EINECS: 213-223-5
UN: 2929
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f5cbc2732a9210dfb896e40a2ecbb47391f57b1&vid=5f5cbc2732a9210dfb896e40a2ecbb47391f57b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR