2-Chloro-4-nitrotoluene ≥99.0%

Supplier: TCI America

C0690-500G C0690-25G
TCC0690-500GEA 170.5 USD
TCC0690-500G TCC0690-25G
2-Chloro-4-nitrotoluene ≥99.0%
2-Chloro-4-nitrotoluene

CAS Number: 121-86-8
MDL Number: MFCD00007210
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 260
Melting point (°C): 63

Formula: C₇H₆ClNO₂
Boiling Pt: 260 °C
Melting Pt: 63 °C
MDL Number: MFCD00007210
CAS Number: 121-86-8
EINECS: 204-501-7
UN: 3457
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2aba2df7400a85e348ca0dc8d1dbfe51249e126&vid=4f9363ac02e64d144885e1cb2b37e3d60c007c58&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR