Cupferron ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Ammonium 2-oxo-1-phenylhydrazinolate
C0646-500G
C0646-25G
TCC0646-500GEA
337
USD
TCC0646-500G
TCC0646-25G
Cupferron ≥98.0% (by HPLC, titration analysis)
Cupferron
CAS Number: 135-20-6
MDL Number: MFCD00078422
Molecular Formula: C6H6N2O2
Molecular Weight: 155.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 164
Lambda max.: 282 nm (H2O)Storage Temperature: 0-10°C
Other Product Names / Synonyms: Ammonium Nitrosophenylhydroxylamine, N-Nitroso-N-phenylhydroxylamine Ammonium Salt
Formula:
C₆H₉N₃O₂ MW: 155.16 Melting Pt: 164 °C |
MDL Number:
MFCD00078422 CAS Number: 135-20-6 EINECS: 205-183-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...