(Chloromethyl)dimethylchlorosilane ≥98.0%

Supplier: TCI America

Synonyms: CMDMCS, Chloro(chloromethyl)dimethylsilane

C0605-25G C0605-250G
TCC0605-025GEA 43 USD
TCC0605-025G TCC0605-250G
(Chloromethyl)dimethylchlorosilane ≥98.0%
(Chloromethyl)dimethylchlorosilane

CAS Number: 1719-57-9
MDL Number: MFCD00000875
Molecular Formula: C3H8Cl2Si
Molecular Weight: 143.08
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 22
Specific Gravity (20/20): 1.09


Product Categories: Materials Chemistry, Silane Coupling Agents, Analytical Science, Silylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: (Chloromethyl)dimethylchlorosilane, CMDMCS

Formula: C₃H₈Cl₂Si
MW: 143.09 g/mol
Boiling Pt: 115 °C
Density: 1.09 (20 °C)
Flash Pt: 22 °C
MDL Number: MFCD00000875
CAS Number: 1719-57-9
EINECS: 217-006-6
UN: 2985
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8b1ee3a9725040e325a90326fcf9525540db7a1&vid=a8b1ee3a9725040e325a90326fcf9525540db7a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR