Cyclohexylméthanol ≥98.0%
Supplier: TCI America
C0595-500G
C0595-25G
C0595-100G
TCC0595-500GEA
330
USD
TCC0595-500G
TCC0595-025G
TCC0595-100G
Cyclohexylméthanol ≥98.0%
Cyclohexylméthanol
CAS Number: 100-49-2
MDL Number: MFCD00001510
Molecular Formula: C7H14O
Molecular Weight: 114.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 183
Flash Point (°C): 71
Specific Gravity (20/20): 0.93
Formula:
C₇H₁₄O Boiling Pt: 183 °C Melting Pt: –43 °C Density: 0.93 (20 °C) Flash Pt: 71 °C |
MDL Number:
MFCD00001510 CAS Number: 100-49-2 EINECS: 202-857-8 Merck Index: 13,02760 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...