Cyclopentanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C0512-5G C0512-25G
TCC0512-005GEA 18.32 USD
TCC0512-005G TCC0512-025G
Cyclopentanecarboxylic acid ≥98.0% (by GC, titration analysis)
Cyclopentanecarboxylic acid

CAS Number: 3400-45-1
MDL Number: MFCD00001371
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 216
Flash Point (°C): 94
Freezing point (°C): -9
Specific Gravity (20/20): 1.05

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 216 °C
Melting Pt: 4 °C
Density: 1.05 (20 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00001371
CAS Number: 3400-45-1
EINECS: 222-269-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd651b2a3a5616f6230c76a47c8028145218a28e&vid=a9b8b3c92b24092fb9a4509a4f3d6639632d7dff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR