Cyclopentanol ≥99.0%
Supplier: TCI America
C0509-25ML
C0509-100ML
TCC0509-25MLEA
16.28
USD
TCC0509-25ML
TCC0509-100ML
Cyclopentanol ≥99.0%
Cyclopentanol
CAS Number: 96-41-3
MDL Number: MFCD00001363
Molecular Formula: C5H10O
Molecular Weight: 86.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Melting point (°C): -19
Flash Point (°C): 47
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: Hydroxycyclopentane
Formula:
C₅H₁₀O MW: 86.13 g/mol Boiling Pt: 140 °C Melting Pt: –19 °C Density: 0.95 (20 °C) Flash Pt: 47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001363 CAS Number: 96-41-3 EINECS: 202-504-8 UN: 2244 ADR: 3,III Merck Index: 13,02770 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...