Cyclohexylamine ≥99.0%

Supplier: TCI America
Danger

Synonyms: Aminocyclohexane

C0494-500ML C0494-25ML
TCC0494-500MLEA 53.63 USD
TCC0494-500ML TCC0494-25ML
Cyclohexylamine ≥99.0%
Cyclohexylamine

CAS Number: 108-91-8
MDL Number: MFCD00001486
Molecular Formula: C6H13N
Molecular Weight: 99.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Melting point (°C): 18
Flash Point (°C): 30
Specific Gravity (20/20): 0.87

Formula: C₆H₁₃N
MW: 99.18 g/mol
Boiling Pt: 135 °C
Melting Pt: 18 °C
Density: 0.87 (20 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00001486
CAS Number: 108-91-8
EINECS: 203-629-0
UN: 2357
ADR: 8,II
Merck Index: 13,02758
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f9058cc95d1295ebb63b097c80d094732a68da&vid=16f9058cc95d1295ebb63b097c80d094732a68da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR