Cyclohexene ≥99.0%
Supplier: TCI America
|
Danger
|
C0491-500ML
C0491-25ML
TCC0491-500MLEA
114.73
USD
TCC0491-500ML
TCC0491-25ML
Cyclohexene ≥99.0%
Cyclohexene
CAS Number: 110-83-8
MDL Number: MFCD00001539
Molecular Formula: C6H10
Molecular Weight: 82.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Melting point (°C): -104
Flash Point (°C): -6
Specific Gravity (20/20): 0.81
Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: 1,2,3,4-Tetrahydrobenzene
Formula:
C₆H₁₀ MW: 82.15 g/mol Boiling Pt: 82 °C Melting Pt: -104 °C Density: 0.81 (20 °C) Flash Pt: -6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001539 CAS Number: 110-83-8 EINECS: 203-807-8 UN: 2256 ADR: 3,II Merck Index: 13,02756 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...