Cyclohexane ≥99.5%
Supplier: TCI America
|
Danger
|
C0469-500ML
C0469-25ML
TCC0469-500MLEA
34.92
USD
TCC0469-500ML
TCC0469-25ML
Cyclohexane ≥99.5%
Cyclohexane
CAS Number: 110-82-7
MDL Number: MFCD00003814
Molecular Formula: C6H12
Molecular Weight: 84.16
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 81
Flash Point (°C): -20
Freezing point (°C): 6
Specific Gravity (20/20): 0.78
Formula:
C₆H₁₂ Boiling Pt: 81 °C Melting Pt: 7 °C Density: 0.78 (20 °C) Flash Pt: -20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003814 CAS Number: 110-82-7 EINECS: 203-806-2 UN: 1145 ADR: 3,II Merck Index: 13,02752 |
|
Specification Test Results
Purity/Analysis Method | >99.5% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...