Cycloheptanone ≥98.0%

Supplier: TCI America

Synonyms: Suberon, Cycloheptan-1-one

C0466-500ML C0466-25ML C0466-100ML
TCC0466-500MLEA 653 USD
TCC0466-500ML TCC0466-25ML TCC0466-100ML
Cycloheptanone ≥98.0%
Cycloheptanone

CAS Number: 502-42-1
MDL Number: MFCD00004159
Molecular Formula: C7H12O
Molecular Weight: 112.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Melting point (°C): -21
Flash Point (°C): 56
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: Suberone

Formula: C₇H₁₂O
MW: 112.17 g/mol
Boiling Pt: 180 °C
Melting Pt: –21 °C
Density: 0.95 (20 °C)
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00004159
CAS Number: 502-42-1
EINECS: 207-937-6
UN: 1224
ADR: 3,III
Merck Index: 13,02751
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=704bb2eeeae2eb499107b02b5771df6ea79eddd2&vid=c32f1e586196dd84c15f832f564cc3561ce9c94c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR