Pyridine-2-carbonitrile ≥99.0%

Supplier: TCI America
Warning

Synonyms: 2-Cyanopyridine, 2-Pyridinecarbonitrile, Picolinonitrile

C0455-25G C0455-500G
TCC0455-025GEA 22 USD
TCC0455-025G TCC0455-500G
Pyridine-2-carbonitrile ≥99.0%
Pyridine-2-carbonitrile

CAS Number: 100-70-9
MDL Number: MFCD00006218
Molecular Formula: C6H4N2
Molecular Weight: 104.11
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 225
Flash Point (°C): 89
Freezing point (°C): 26


Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Picolinonitrile, 2-Pyridinecarbonitrile

Formula: C₆H₄N₂
MW: 104.11 g/mol
Boiling Pt: 212
Melting Pt: 26…28 °C
Density: 1.081 g/cm³ (25 °C)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00006218
CAS Number: 100-70-9
EINECS: 202-880-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50f12477d645d64a1527dc7d983ef822a026e5aa&vid=50f12477d645d64a1527dc7d983ef822a026e5aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR