Cyanoguanidine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 461-58-5
MDL Number: MFCD00008066
Molecular Formula: C2H4N4
Molecular Weight: 84.08
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 211
Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: Cyanoguanidine
Formula:
(H₂N)₂CNCN Boiling Pt: 143 °C (1013 hPa) Melting Pt: 211 °C Density: 1.404 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008066 CAS Number: 461-58-5 EINECS: 207-312-8 Merck Index: 13,03119 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...