3-Cyanobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C0446-25G C0446-5G
TCC0446-025GEA 167.4 USD
TCC0446-025G TCC0446-5G
3-Cyanobenzoic acid ≥98.0% (by GC, titration analysis)
3-Cyanobenzoic acid

CAS Number: 1877-72-1
MDL Number: MFCD00002486
Molecular Formula: C8H5NO2
Molecular Weight: 147.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 220

Formula: C₈H₅NO₂
MW: 147.13 g/mol
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00002486
CAS Number: 1877-72-1
EINECS: 217-511-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e1650b51ae0619ff98b336821cb27c24128a12c&vid=2e6fea72c18912cf747c25880380e93c51605ba4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR