4-Cyanobenzaldehyde ≥98.0%

Supplier: TCI America

Synonyms: 4-Formylbenzonitrile

C0443-25G C0443-5G
TCC0443-025GEA 53.13 USD
TCC0443-025G TCC0443-5G
4-Cyanobenzaldehyde ≥98.0%
4-Cyanobenzaldehyde

CAS Number: 105-07-7
MDL Number: MFCD00003376
Molecular Formula: C8H5NO
Molecular Weight: 131.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 133
Melting point (°C): 100


Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction), Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Cyanobenzaldehyde

Formula: C₈H₅NO
MW: 131.13 g/mol
Boiling Pt: 133 °C
Melting Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003376
CAS Number: 105-07-7
EINECS: 203-267-3
UN: 3439
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3250afff6039a43ab4d6c49711d09e214d38cafb&vid=3250afff6039a43ab4d6c49711d09e214d38cafb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR