Cyanoacetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

C0439-500G C0439-25G
TCC0439-500GEA 258 USD
TCC0439-500G TCC0439-25G
Cyanoacetic acid ≥98.0% (by titrimetric analysis)
Cyanoacetic acid

CAS Number: 372-09-8
MDL Number: MFCD00002677
Molecular Formula: C3H3NO2
Molecular Weight: 85.06
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 108
Melting point (°C): 71
Flash Point (°C): 107

Formula: C₃H₃NO₂
MW: 85.06 g/mol
Boiling Pt: 108 °C
Melting Pt: 71 °C
Density: Min. 1.1 g/cm³ (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00002677
CAS Number: 372-09-8
EINECS: 206-743-9
UN: 3261
ADR: 8,II
Merck Index: 13,02718
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ef2d6aba01aeba29a29e2fb2de3de743d556e3d&vid=5ef2d6aba01aeba29a29e2fb2de3de743d556e3d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR