3-Cyanobenzaldehyde ≥98.0%

Supplier: TCI America
Warning

Synonyms: 3-Formylbenzonitrile

C0429-5G C0429-25G
TCC0429-005GEA 88.53 USD
TCC0429-005G TCC0429-025G
3-Cyanobenzaldehyde ≥98.0%
3-Cyanobenzaldehyde

CAS Number: 24964-64-5
MDL Number: MFCD00003344
Molecular Formula: C8H5NO
Molecular Weight: 131.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 210
Melting point (°C): 79


Other Product Names / Synonyms: 3-Formylbenzonitrile

Formula: C₈H₅NO
MW: 131.13 g/mol
Boiling Pt: 210 °C
Melting Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00003344
CAS Number: 24964-64-5
EINECS: 246-549-1
UN: 2811
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f87b1e2be6f9ac65a9c3635250b6f807ec49a55&vid=4f87b1e2be6f9ac65a9c3635250b6f807ec49a55&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR