Cresol (mixture of isomers)

Supplier: TCI America
Danger

C0412-25G C0412-500G
TCC0412-25GEA 17 USD
TCC0412-25G TCC0412-500G
Cresol (mixture of isomers)
Cresol (mixture of isomers)

CAS Number: 1319-77-3
MDL Number: MFCD00151099
Molecular Formula: C7H8O
Molecular Weight: 108.14
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 192
Flash Point (°C): 81
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: Cresylic Acid, Tricresol

Formula: C₇H₈O
Boiling Pt: 192 °C
Density: 1.03 (20 °C)
Flash Pt: 81 °C
MDL Number: MFCD00151099
CAS Number: 1319-77-3
EINECS: 215-293-2
Merck Index: 12,02645
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3c0123545f1c38fc298abd2efe9e7da23f46ffb&vid=d3c0123545f1c38fc298abd2efe9e7da23f46ffb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR