2-Hydroxy-p-toluic acid ≥99.0% (by HPLC)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Hydroxy-4-methylbenzoic acid, m-Homosalicyclic acid, m-Cresotic acid, 4-Methylsalicylic acid
C0409-25G
C0409-500G
TCC0409-025GEA
26.46
USD
TCC0409-025G
TCC0409-500G
2-Hydroxy-p-toluic acid ≥99.0% (by HPLC)
2-Hydroxy-p-toluic acid
CAS Number: 50-85-1
MDL Number: MFCD00002454
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 177
Other Product Names / Synonyms: m-Cresotic Acid, 2-Hydroxy-4-methylbenzoic Acid, 2-Hydroxy-p-toluic Acid
Formula:
C₈H₈O₃ MW: 152.15 g/mol Melting Pt: 177 °C |
MDL Number:
MFCD00002454 CAS Number: 50-85-1 EINECS: 200-068-3 |
Specification Test Results
Purity/Analysis Method | >99.0% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...