m-Cresol ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
1-Hydroxy-3-methylbenzene, 3-Methylphenol, 3-Hydroxytoluol, 3-Hydroxytoluene
C0401-500G
C0401-25G
TCC0401-500GEA
69.88
USD
TCC0401-500G
TCC0401-25G
m-Cresol ≥98.0%
m-Cresol
CAS Number: 108-39-4
MDL Number: MFCD00002302
Molecular Formula: C7H8O
Molecular Weight: 108.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 202
Flash Point (°C): 86
Freezing point (°C): 10
Specific Gravity (20/20): 1.04
Lambda max.: 273 nm
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis
Other Product Names / Synonyms: 3-Methylphenol
Formula:
CH₃C₆H₄OH MW: 108.14 g/mol Boiling Pt: 202 °C Melting Pt: 11.5 °C Density: 1.04 (20 °C) Flash Pt: 86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002302 CAS Number: 108-39-4 EINECS: 203-577-9 UN: 2076 ADR: 6.1,II Merck Index: 13,02604 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...