p-Cresol ≥99.0%
Supplier: TCI America
|
Danger
|
CAS Number: 106-44-5
MDL Number: MFCD00002376
Molecular Formula: C7H8O
Molecular Weight: 108.14
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 202
Flash Point (°C): 93
Freezing point (°C): 32
Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Methylphenol
Formula:
C₇H₈O Boiling Pt: 202 °C Melting Pt: 32 - 34 °C Density: 1.034 g/cm³ (20 °C) Flash Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002376 CAS Number: 106-44-5 EINECS: 203-398-6 UN: 3455 ADR: 6.1,II Merck Index: 13,02604 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...