(+)-Cinchonine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Cinchonine, (S)-Quinolin-4-yl((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol

C0350-25G C0350-200G
TCC0350-025GEA 67.16 USD
TCC0350-025G TCC0350-200G
(+)-Cinchonine ≥98.0% (by titrimetric analysis)
(+)-Cinchonine

CAS Number: 118-10-5
MDL Number: MFCD00064372
Molecular Formula: C19H22N2O
Molecular Weight: 294.40
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 261
Specific rotation [a]20/D: 223 deg (C=0.5, EtOH)

Formula: C₁₉H₂₂N₂O
MW: 294.4 g/mol
Melting Pt: 261 °C
Density: 1.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064372
CAS Number: 118-10-5
EINECS: 204-234-6
Merck Index: 13,02308
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5ad1624662545d0483e767665bf4ed92bee3f83&vid=b5ad1624662545d0483e767665bf4ed92bee3f83&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR