(-)-Cinchonidine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (8α,9R)-Cinchonan-9-ol

C0347-25G C0347-100G
TCC0347-025GEA 70 USD
TCC0347-025G TCC0347-100G
(-)-Cinchonidine ≥98.0% (by titrimetric analysis)
(-)-Cinchonidine

CAS Number: 485-71-2
MDL Number: MFCD00006783
Molecular Formula: C19H22N2O
Molecular Weight: 294.40
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 206
Specific rotation [a]20/D: -108 deg (C=1, EtOH)

Formula: C₁₉H₂₂N₂O
MW: 294.4 g/mol
Boiling Pt: 465 °C (1013 hPa)
Melting Pt: 206 °C
Density: 1.22 g/cm³ (20 °C)
MDL Number: MFCD00006783
CAS Number: 485-71-2
EINECS: 207-622-3
Merck Index: 13,02307
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d47e7949f8fff486ed24cbeb76dd5cc44cf9bc3b&vid=d47e7949f8fff486ed24cbeb76dd5cc44cf9bc3b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR