α-Chloro-p-xylene ≥98.0%

Supplier: TCI America
Warning

Synonyms: alpha-Chloro-p-xylene, 4-Methylbenzyl chloride

C0312-500G C0312-25G
TCC0312-500GEA 97 USD
TCC0312-500G TCC0312-25G
α-Chloro-p-xylene ≥98.0%
α-Chloro-p-xylene

CAS Number: 104-82-5
MDL Number: MFCD00000919
Molecular Formula: C8H9Cl
Molecular Weight: 140.61
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 202
Flash Point (°C): 97
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: 4-Methylbenzyl Chloride

Formula: C₈H₉Cl
MW: 140.61 g/mol
Boiling Pt: 202 °C
Melting Pt: 5 °C
Density: 1.07 (20 °C)
Flash Pt: 97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000919
CAS Number: 104-82-5
EINECS: 203-241-1
UN: 3265
ADR: 8,II
Merck Index: 12,10224
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed63e7885d222b58abc168c7f9b12b55ed14783a&vid=446dc86687526998c6a216fb02db21fd8d6a716b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR